Assay Details
Target Gene Details
Entrez Gene ID: | 131831 |
Gene Name: | glutamate rich 6 |
Gene Aliases: |
C3orf44, ERICH6A, FAM194A |
Location: |
Chr.3:150659885-150704563 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH6 | NM_001308234.1 | 14 | 1663 | NP_001295163.1 |
| NM_152394.4 | 14 | 1967 | NP_689607.2 | |
| XM_005247120.3 | XP_005247177.1 | |||
| BC029577.1 | 14 | 1647 | AAH29577.1 | |
| BC036239.1 | 14 | 1951 | AAH36239.2 |

Genomic Map