Assay Details
Target Gene Details
Entrez Gene ID: | 8807 |
Gene Name: | interleukin 18 receptor accessory protein |
Gene Aliases: |
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB |
Location: |
Chr.2:102418551-102452568 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18RAP | NM_003853.3 | 6 | 1126 | NP_003844.1 |
| XM_011512087.2 | 6 | 900 | XP_011510389.1 | |
| XM_011512088.2 | 3 | 348 | XP_011510390.1 | |
| AF077346.1 | 6 | 1121 | AAC72196.1 | |
| AK300026.1 | ||||
| BC069630.1 | 4 | 723 | AAH69630.1 | |
| BC106764.2 | 4 | 525 | AAI06765.1 | |
| BC106765.2 | 5 | 850 | AAI06766.1 | |
| BC137474.1 | 4 | 666 | ||
| BC137475.1 | 4 | 666 |
More Information
Set Membership: |
|

Genomic Map