Assay Details
Target Gene Details
Entrez Gene ID: | 23362 |
Gene Name: | pleckstrin and Sec7 domain containing 3 |
Gene Aliases: |
EFA6D, EFA6R, HCA67 |
Location: |
Chr.8:18527303-19086909 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSD3 | NM_015310.3 | 16 | 8015 | NP_056125.3 |
| NM_206909.2 | 13 | 6419 | NP_996792.1 | |
| XM_017013258.1 | 16 | 9324 | XP_016868747.1 | |
| XM_017013259.1 | 16 | 9302 | XP_016868748.1 | |
| XM_017013260.1 | 16 | 8075 | XP_016868749.1 | |
| XM_017013261.1 | 15 | 8289 | XP_016868750.1 | |
| XM_017013265.1 | 13 | 6825 | XP_016868754.1 | |
| AB023159.2 | 13 | 6400 | ||
| AF243495.2 | 14 | 6425 | AAF61269.2 | |
| AL832778.1 | 15 | 7882 | ||
| BX647320.1 | 6 | 5722 | CAI46004.1 | |
| BX647406.1 | 8 | 6066 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428196 | Chr.8:18219826 - 18542377 on Build GRCh38 | Gain |
|
| nsv831252 | Chr.8:18347493 - 18548287 on Build GRCh38 | Gain |
|
| nsv1026091 | Chr.8:18441326 - 18617605 on Build GRCh38 | Gain |
|
| nsv1026366 | Chr.8:18071966 - 18557180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map