Assay Details
Target Gene Details
Entrez Gene ID: | 832 |
Gene Name: | capping actin protein of muscle Z-line beta subunit |
Gene Aliases: |
CAPB, CAPPB, CAPZ |
Location: |
Chr.1:19338773-19485641 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZB | NM_001206540.2 | 10 | 1276 | NP_001193469.1 |
| NM_001206541.2 | 9 | 1583 | NP_001193470.1 | |
| NM_001282162.1 | 9 | 1475 | NP_001269091.1 | |
| NM_004930.4 | 9 | 1163 | NP_004921.1 | |
| XM_006710938.3 | 10 | 1169 | XP_006711001.1 | |
| XM_011542228.2 | 10 | 2097 | XP_011540530.1 | |
| XM_011542229.2 | 9 | 1284 | XP_011540531.1 | |
| XM_011542230.2 | 9 | 1259 | XP_011540532.1 | |
| XM_017002428.1 | 10 | 1141 | XP_016857917.1 | |
| AI799520.1 | 1 | 33 | ||
| AK022336.1 | 1 | 1152 | ||
| AK125085.1 | 9 | 1455 | ||
| AK126650.1 | 1 | 4696 | ||
| AK301442.1 | 9 | 1477 | ||
| BC008095.1 | 3 | 386 | ||
| BC024601.1 | 6 | 712 | AAH24601.1 | |
| BC107752.1 | AAI07753.1 | |||
| BQ009850.1 | 1 | 628 | ||
| BQ773230.1 | ||||
| U03271.1 | 8 | 960 | AAA87395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map