Assay Details
Target Gene Details
Entrez Gene ID: | 80975 |
Gene Name: | transmembrane protease, serine 5 |
Gene Aliases: |
SPINESIN |
Location: |
Chr.11:113687546-113709811 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS5 | NM_001288749.1 | 10 | 1602 | NP_001275678.1 |
| NM_001288750.1 | 12 | 1809 | NP_001275679.1 | |
| NM_001288751.1 | 13 | 2016 | NP_001275680.1 | |
| NM_001288752.1 | 11 | 1705 | NP_001275681.1 | |
| NM_030770.3 | 13 | 1912 | NP_110397.2 | |
| NR_110046.1 | 11 | 1710 | ||
| NR_110047.1 | 10 | 1557 | ||
| XM_017018366.1 | 10 | 6376 | XP_016873855.1 | |
| XM_017018367.1 | 12 | 6568 | XP_016873856.1 | |
| AB028140.1 | 13 | 1918 | BAB20375.1 | |
| AI056603.1 | 1 | 349 | ||
| BC121802.1 | 12 | 1782 | ||
| BC121803.1 | 13 | 1989 |
More Information
Additional Information:
For this assay, SNP(s) [rs146829461] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map