Assay Details
Target Gene Details
Entrez Gene ID: | 285761 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
dJ94G16.1 |
Location: |
Chr.6:117480346-117569858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCBLD1 | NM_173674.2 | NP_775945.1 | ||
| BC035671.1 | AAH35671.1 | |||
| CR749448.1 | CAH18285.1 |
Target Gene Details
Entrez Gene ID: | 57120 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
CAL, FIG, GOPC1, PIST, dJ94G16.2 |
Location: |
Chr.6:117560269-117602542 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOPC | NM_001017408.2 | 6 | 1244 | NP_001017408.1 |
| NM_020399.3 | 7 | 1268 | NP_065132.1 | |
| AB209385.1 | 6 | 2337 | BAD92622.1 | |
| AF287894.1 | 7 | 1090 | AAG00572.1 | |
| AF450008.1 | 6 | 1232 | AAL47160.1 | |
| AK314486.1 | 6 | 1215 | ||
| AY033606.1 | 6 | 1197 | AAK57733.1 | |
| BC009553.2 | 6 | 1179 | AAH09553.1 | |
| KF420122.1 | ||||
| KF420123.1 |
More Information
Set Membership: |
|

Genomic Map