Assay Details
Target Gene Details
Entrez Gene ID: | 2328 |
Gene Name: | flavin containing monooxygenase 3 |
Gene Aliases: |
FMOII, TMAU, dJ127D3.1 |
Location: |
Chr.1:171090873-171117819 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMO3 | NM_001002294.2 | 3 | 342 | NP_001002294.1 |
| NM_001319173.1 | 4 | 452 | NP_001306102.1 | |
| NM_001319174.1 | NP_001306103.1 | |||
| NM_006894.5 | 3 | 325 | NP_008825.4 | |
| XM_011509345.2 | 4 | 473 | XP_011507647.1 | |
| AK223166.1 | 3 | 334 | BAD96886.1 | |
| AK296105.1 | ||||
| AK298406.1 | 4 | 437 | ||
| AK313197.1 | 3 | 315 | ||
| BC032016.1 | 3 | 277 | AAH32016.1 | |
| BM453951.1 | 3 | 354 | ||
| KU178077.1 | 2 | 231 | ||
| KU178078.1 | ||||
| M83772.1 | 3 | 367 | AAA86284.1 | |
| Z47552.1 | 3 | 324 | CAA87632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map