Assay Details
Target Gene Details
Entrez Gene ID: | 81027 |
Gene Name: | tubulin beta 1 class VI |
Gene Aliases: |
- |
Location: |
Chr.20:59016026-59026654 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB1 | NM_030773.3 | 4 | 3294 | NP_110400.1 |
| XM_017028085.1 | 4 | 3488 | XP_016883574.1 | |
| AJ292757.1 | 4 | 3294 | CAC16605.1 | |
| BC033679.1 | 4 | 3127 | AAH33679.1 |
More Information
Set Membership: |
|

Genomic Map