Assay Details
Target Gene Details
Entrez Gene ID: | 259232 |
Gene Name: | sodium leak channel, non-selective |
Gene Aliases: |
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1 |
Location: |
Chr.13:101053774-101417206 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN | NM_052867.2 | 40 | 4685 | NP_443099.1 |
| XM_011521067.2 | 40 | 4625 | XP_011519369.1 | |
| XM_011521068.2 | 40 | 4608 | XP_011519370.1 | |
| XM_011521069.2 | 39 | 4538 | XP_011519371.1 | |
| XM_017020534.1 | 39 | 4569 | XP_016876023.1 | |
| XM_017020535.1 | 39 | 4511 | XP_016876024.1 | |
| XM_017020536.1 | 35 | 4437 | XP_016876025.1 | |
| XM_017020537.1 | 34 | 3826 | XP_016876026.1 | |
| AK055506.1 | 7 | 742 | ||
| AK126621.1 | 21 | 2149 | ||
| AK126718.1 | 13 | 1286 | ||
| AK172752.1 | 3 | 216 | BAD18738.1 | |
| AY141972.1 | 40 | 4579 | AAN10255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1149 | Chr.13:101042454 - 101087249 on Build GRCh38 | Deletion |
|
| nsv832699 | Chr.13:100951779 - 101142880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map