Assay Details
Target Gene Details
Entrez Gene ID: | 405753 |
Gene Name: | dual oxidase maturation factor 2 |
Gene Aliases: |
SIMNIPHOM, TDH5 |
Location: |
Chr.15:45114321-45118106 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUOXA2 | NM_207581.3 | 4 | 658 | NP_997464.2 |
| XM_017022180.1 | 4 | 713 | XP_016877669.1 | |
| AK303955.1 | 4 | 657 | ||
| BC137465.1 | 4 | 579 | ||
| BX537581.1 | 4 | 942 | ||
| DQ489734.1 | 4 | 509 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map