Assay Details
Target Gene Details
Entrez Gene ID: | 23107 |
Gene Name: | mitochondrial ribosomal protein S27 |
Gene Aliases: |
MRP-S27, S27mt |
Location: |
Chr.5:72219409-72320257 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS27 | NM_001286748.1 | 9 | 736 | NP_001273677.1 |
| NM_001286751.1 | 8 | 662 | NP_001273680.1 | |
| NM_015084.2 | 8 | 694 | NP_055899.2 | |
| AK025417.1 | 8 | 677 | ||
| AK027001.1 | 1 | 66 | ||
| AK128683.1 | 1 | 1252 | ||
| AK299412.1 | 9 | 710 | ||
| AK300111.1 | 8 | 660 | ||
| AK309065.1 | 8 | 644 | ||
| BC003632.2 | 2 | 84 | AAH03632.1 | |
| BC030521.2 | 2 | 148 | AAH30521.1 | |
| BC064902.1 | 8 | 665 | AAH64902.1 | |
| D87453.1 | 8 | 663 |
More Information
Set Membership: |
|

Genomic Map