Assay Details
Target Gene Details
Entrez Gene ID: | 1010 |
Gene Name: | cadherin 12 |
Gene Aliases: |
CDHB |
Location: |
Chr.5:21750191-22853622 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH12 | NM_001317227.1 | 8 | 1780 | NP_001304156.1 |
| NM_001317228.1 | 8 | 1775 | NP_001304157.1 | |
| NM_004061.4 | 11 | 2409 | NP_004052.2 | |
| XM_011513927.2 | 9 | 1686 | XP_011512229.1 | |
| XM_017008920.1 | 10 | 2036 | XP_016864409.1 | |
| XM_017008921.1 | 11 | 2129 | XP_016864410.1 | |
| AK092176.1 | 6 | 1059 | ||
| AK295123.1 | 8 | 1775 | ||
| AK314800.1 | 8 | 1780 | ||
| BC047608.1 | 11 | 2114 | AAH47608.1 | |
| L33477.1 | 11 | 2410 | AAB48539.1 | |
| L34057.1 | 7 | 1415 | AAA35623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428115 | Chr.5:21778632 - 22090409 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map