Assay Details
Target Gene Details
Entrez Gene ID: | 25980 |
Gene Name: | AAR2 splicing factor homolog |
Gene Aliases: |
C20orf4, CGI-23 |
Location: |
Chr.20:36236136-36256941 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAR2 | NM_001271874.1 | 2 | 580 | NP_001258803.1 |
| NM_015511.4 | 2 | 859 | NP_056326.2 | |
| XM_006723770.3 | 3 | 1296 | XP_006723833.1 | |
| XM_011528762.2 | 3 | 670 | XP_011527064.1 | |
| XM_011528763.2 | 3 | 939 | XP_011527065.1 | |
| AF132957.1 | 1 | 500 | AAD27732.1 | |
| AL117419.1 | 2 | 520 | CAB55913.1 | |
| BC001751.1 | 1 | 507 | AAH01751.1 | |
| BC019311.1 | 2 | 751 | AAH19311.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833965 | Chr.20:36085697 - 36284604 on Build GRCh38 | Loss |
|
| nsv1066673 | Chr.20:35840156 - 36398197 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map