Assay Details
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | NP_443728.3 | ||
| XM_006713487.3 | XP_006713550.1 | |||
| XM_011512390.2 | XP_011510692.1 | |||
| XM_011512394.2 | XP_011510696.1 | |||
| XM_011512399.2 | XP_011510701.1 | |||
| XM_017005676.1 | XP_016861165.1 | |||
| XM_017005677.1 | XP_016861166.1 | |||
| XM_017005678.1 | XP_016861167.1 | |||
| XM_017005679.1 | XP_016861168.1 | |||
| XM_017005680.1 | XP_016861169.1 | |||
| AB046855.1 | BAB13461.1 | |||
| AF399708.1 | AAO38813.1 | |||
| AK128165.1 | ||||
| BC144559.1 |
Target Gene Details
Entrez Gene ID: | 64805 |
Gene Name: | purinergic receptor P2Y12 |
Gene Aliases: |
ADPG-R, BDPLT8, HORK3, P2T(AC), P2Y(12)R, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12, SP1999 |
Location: |
Chr.3:151336843-151384812 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY12 | NM_022788.4 | 3 | 645 | NP_073625.1 |
| NM_176876.2 | 2 | 619 | NP_795345.1 | |
| XM_017007069.1 | 3 | 903 | XP_016862558.1 | |
| AB052684.1 | 1 | 345 | BAB60824.1 | |
| AF313449.1 | 2 | 419 | AAG48944.1 | |
| AF321815.1 | 2 | 416 | AAK00948.1 | |
| AJ320495.1 | 1 | 362 | CAC87144.1 | |
| AK292096.1 | 3 | 584 | ||
| AK303714.1 | ||||
| AK316575.1 | 3 | 587 | ||
| AY136754.1 | 1 | 345 | AAN01280.1 | |
| BC017898.1 | 3 | 589 | AAH17898.1 | |
| DA267276.1 | 2 | 434 | ||
| DA343254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829757 | Chr.3:151327092 - 151508654 on Build GRCh38 | Loss |
|
| nsv954492 | Chr.3:151315313 - 151363512 on Build GRCh38 | Duplication |
|
| nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map