Assay Details
Target Gene Details
Entrez Gene ID: | 55753 |
Gene Name: | oxoglutarate dehydrogenase-like |
Gene Aliases: |
- |
Location: |
Chr.10:49734641-49762379 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGDHL | NM_001143996.1 | NP_001137468.1 | ||
| NM_001143997.1 | NP_001137469.1 | |||
| NM_018245.2 | NP_060715.2 | |||
| XM_011539946.1 | XP_011538248.1 | |||
| XM_011539947.1 | XP_011538249.1 | |||
| XM_017016401.1 | XP_016871890.1 | |||
| XM_017016402.1 | XP_016871891.1 | |||
| XM_017016403.1 | XP_016871892.1 | |||
| AB033116.3 | ||||
| AK001713.1 | BAA91855.1 | |||
| AK055817.1 | ||||
| AK225558.1 | ||||
| AK290226.1 | ||||
| AK296551.1 | ||||
| AK303729.1 | ||||
| BC026320.1 | AAH26320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048905 | Chr.10:49720681 - 49811144 on Build GRCh38 | Gain |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map