Assay Details
Target Gene Details
Entrez Gene ID: | 220965 |
Gene Name: | family with sequence similarity 13 member C |
Gene Aliases: |
FAM13C1 |
Location: |
Chr.10:59245933-59363186 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM13C | XM_005269618.4 | 14 | 3469 | XP_005269675.2 |
XM_005269619.4 | 12 | 3175 | XP_005269676.2 | |
XM_006717702.3 | 16 | 3473 | XP_006717765.1 | |
XM_006717703.3 | 16 | 3655 | XP_006717766.1 | |
XM_011539490.2 | 15 | 3528 | XP_011537792.1 | |
XM_011539491.2 | 16 | 3574 | XP_011537793.1 | |
XM_017015885.1 | 15 | 3386 | XP_016871374.1 | |
XM_017015886.1 | 15 | 3640 | XP_016871375.1 | |
XM_017015887.1 | 15 | 3390 | XP_016871376.1 | |
XM_017015888.1 | 16 | 3652 | XP_016871377.1 | |
XM_017015889.1 | 16 | 3470 | XP_016871378.1 | |
XM_017015890.1 | 16 | 3709 | XP_016871379.1 | |
XM_017015891.1 | 13 | 3349 | XP_016871380.1 | |
XM_017015892.1 | 13 | 3346 | XP_016871381.1 | |
XM_017015893.1 | 15 | 3450 | XP_016871382.1 |
Target Gene Details
Entrez Gene ID: | 84457 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein like |
Gene Aliases: |
- |
Location: |
Chr.10:59174811-59247774 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYHIPL | NM_001143774.1 | 5 | 1620 | NP_001137246.1 |
NM_032439.3 | 5 | 1840 | NP_115815.2 | |
XM_011540275.2 | 6 | 1902 | XP_011538577.1 | |
XM_011540276.2 | 5 | 2733 | XP_011538578.1 | |
XM_017016782.1 | 6 | 1721 | XP_016872271.1 | |
XM_017016783.1 | 6 | 2824 | XP_016872272.1 | |
AB058699.1 | 3 | 1181 | BAB47425.1 | |
AK054956.1 | 5 | 1644 | BAB70832.1 | |
AL365474.1 | 5 | 1620 | CAB97028.1 | |
AL834339.1 | 5 | 1841 | CAD39006.1 | |
BC011268.2 | 5 | 1679 | AAH11268.2 | |
CR749429.1 | 6 | 1869 | CAH18267.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
