Assay Details
Target Gene Details
Entrez Gene ID: | 91 |
Gene Name: | activin A receptor type 1B |
Gene Aliases: |
ACTRIB, ACVRLK4, ALK4, SKR2 |
Location: |
Chr.12:51951667-51997079 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR1B | NM_004302.4 | NP_004293.1 | ||
| NM_020327.3 | NP_064732.3 | |||
| NM_020328.3 | NP_064733.3 | |||
| XM_011538966.2 | XP_011537268.1 | |||
| XM_017020199.1 | XP_016875688.1 | |||
| XM_017020200.1 | XP_016875689.1 | |||
| XM_017020201.1 | XP_016875690.1 | |||
| AK299120.1 | ||||
| AK299496.1 | ||||
| BC000254.1 | AAH00254.1 | |||
| BC040531.1 | AAH40531.1 | |||
| BT007072.1 | AAP35735.1 | |||
| L10125.1 | AAA60555.1 | |||
| L10126.1 | 7 | 1624 | AAA60556.1 | |
| U14722.1 | AAA50246.1 | |||
| Z22536.1 | CAA80258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525457 | Chr.12:51955304 - 51991027 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map