Assay Details
Target Gene Details
Entrez Gene ID: | 6160 |
Gene Name: | ribosomal protein L31 |
Gene Aliases: |
L31 |
Location: |
Chr.2:101002229-101019693 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL31 | NM_001098577.2 | NP_001092047.1 | ||
| AK297167.1 | ||||
| AK297247.1 | ||||
| AK297483.1 | ||||
| BE564858.1 | ||||
| CN269912.1 | ||||
| CN269915.1 |
Target Gene Details
Entrez Gene ID: | 11138 |
Gene Name: | TBC1 domain family member 8 |
Gene Aliases: |
AD3, GRAMD8, HBLP1, TBC1D8A, VRP |
Location: |
Chr.2:101006435-101195326 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D8 | NM_001102426.1 | NP_001095896.1 | ||
| XM_005263862.3 | XP_005263919.1 | |||
| XM_005263863.4 | XP_005263920.1 | |||
| XM_005263864.3 | XP_005263921.1 | |||
| XM_011510507.1 | XP_011508809.1 | |||
| XM_017003198.1 | XP_016858687.1 | |||
| XM_017003199.1 | XP_016858688.1 | |||
| AB017114.1 | BAA76517.1 | |||
| AB449889.1 | ||||
| AK000748.1 | ||||
| AK292826.1 | ||||
| BE842460.1 | ||||
| CD722231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834316 | Chr.2:100916586 - 101115039 on Build GRCh38 | Gain |
|
| esv34123 | Chr.2:101008454 - 101391851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map