Assay Details
Target Gene Details
Entrez Gene ID: | 161823 |
Gene Name: | adenosine deaminase like |
Gene Aliases: |
- |
Location: |
Chr.15:43330351-43354555 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAL | NM_001012969.3 | NP_001012987.1 | ||
| NM_001159280.2 | NP_001152752.1 | |||
| NM_001324364.1 | NP_001311293.1 | |||
| NM_001324365.1 | NP_001311294.1 | |||
| NM_001324366.1 | NP_001311295.1 | |||
| NM_001324367.1 | NP_001311296.1 | |||
| NM_001324368.1 | NP_001311297.1 | |||
| XM_011521291.2 | XP_011519593.1 | |||
| XM_017021964.1 | XP_016877453.1 | |||
| XM_017021965.1 | XP_016877454.1 | |||
| XM_017021966.1 | XP_016877455.1 | |||
| XM_017021967.1 | XP_016877456.1 | |||
| XM_017021968.1 | XP_016877457.1 | |||
| XM_017021969.1 | XP_016877458.1 | |||
| AK298870.1 | ||||
| BC075857.1 | AAH75857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1512 | Chr.15:43324636 - 43369448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map