Assay Details
Target Gene Details
Entrez Gene ID: | 56997 |
Gene Name: | coenzyme Q8A |
Gene Aliases: |
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9 |
Location: |
Chr.1:226939339-226987545 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ8A | NM_020247.4 | 3 | 600 | NP_064632.2 |
| XM_005273201.1 | 3 | 496 | XP_005273258.1 | |
| XM_011544238.1 | 4 | 692 | XP_011542540.1 | |
| XM_011544239.2 | 5 | 715 | XP_011542541.1 | |
| XM_011544240.2 | 3 | 509 | XP_011542542.1 | |
| XM_011544241.2 | 3 | 471 | XP_011542543.1 | |
| XM_017001852.1 | 3 | 690 | XP_016857341.1 | |
| AB073905.1 | 3 | 523 | BAB91363.1 | |
| AK074693.1 | 3 | 543 | BAC11143.1 | |
| AK090494.1 | 3 | 337 | ||
| AK092784.1 | 3 | 323 | ||
| AK126200.1 | ||||
| AK293571.1 | 4 | 497 | ||
| AK297735.1 | ||||
| AK301012.1 | 5 | 521 | ||
| BC005171.2 | 3 | 549 | AAH05171.2 | |
| BC129930.1 | 9 | 1442 | ||
| BX648860.1 | 3 | 421 | CAH56132.1 | |
| DA925261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520923 | Chr.1:226939917 - 226991247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map