Assay Details
Target Gene Details
Entrez Gene ID: | 8811 |
Gene Name: | galanin receptor 2 |
Gene Aliases: |
GAL2-R, GALNR2, GALR-2 |
Location: |
Chr.17:76070822-76077541 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALR2 | XM_011525427.2 | XP_011523729.1 |
Target Gene Details
Entrez Gene ID: | 6730 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
- |
Location: |
Chr.17:76038775-76072526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRP68 | NM_001260502.1 | NP_001247431.1 | ||
| NM_014230.3 | NP_055045.2 | |||
| NR_048541.1 | ||||
| AF195951.1 | AAF24308.1 | |||
| AK002002.1 | ||||
| AK074698.1 | BAC11145.1 | |||
| AK126258.1 | ||||
| AK301100.1 | ||||
| AL512708.1 | ||||
| AU137443.1 | ||||
| BC020238.1 | AAH20238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2142 | Chr.17:76070185 - 76087802 on Build GRCh38 | Insertion |
|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
| nsv509674 | Chr.17:76052197 - 76106436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map