Assay Details
Target Gene Details
Entrez Gene ID: | 54883 |
Gene Name: | CWC25 spliceosome associated protein homolog |
Gene Aliases: |
CCDC49 |
Location: |
Chr.17:38800434-38825350 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWC25 | NM_017748.4 | 7 | 983 | NP_060218.1 |
| NR_073428.1 | 8 | 1100 | ||
| AK000298.1 | 7 | 933 | BAA91065.1 | |
| AK225373.1 | 7 | 953 | ||
| AK296115.1 | 8 | 1065 | ||
| AK302983.1 | 7 | 1076 | ||
| AK316077.1 | 6 | 929 | ||
| BC008833.2 | 7 | 969 | AAH08833.1 | |
| CR749362.1 | CAH18215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv833439 | Chr.17:38743861 - 38923753 on Build GRCh38 | Loss |
|
| esv1005760 | Chr.17:38803490 - 38809759 on Build GRCh38 | Deletion |
|

Genomic Map