Assay Details
Target Gene Details
Entrez Gene ID: | 5156 |
Gene Name: | platelet derived growth factor receptor alpha |
Gene Aliases: |
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA |
Location: |
Chr.4:54229097-54298245 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFRA | NM_006206.4 | 14 | 2282 | NP_006197.1 |
| XM_005265743.1 | 14 | 2066 | XP_005265800.1 | |
| XM_006714039.3 | 15 | 2327 | XP_006714102.1 | |
| XM_006714041.3 | 15 | 2330 | XP_006714104.1 | |
| XM_011534385.2 | 14 | 2320 | XP_011532687.1 | |
| XM_017008280.1 | 14 | 2497 | XP_016863769.1 | |
| XM_017008281.1 | 14 | 2563 | XP_016863770.1 | |
| XM_017008282.1 | 14 | 2211 | XP_016863771.1 | |
| AK311006.1 | 13 | 1769 | ||
| AK316578.1 | 14 | 2084 | ||
| BC063414.1 | 14 | 2066 | AAH63414.1 | |
| L25829.1 | 2 | 376 | ||
| M21574.1 | 14 | 2090 | AAA96715.1 | |
| X76079.1 | 2 | 376 |
More Information
Set Membership: |
|

Genomic Map