Assay Details
Target Gene Details
Entrez Gene ID: | 26059 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
CAST, CAST1, ELKSL, SPBC110, Spc110 |
Location: |
Chr.3:55508311-56468473 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERC2 | NM_015576.2 | 18 | 5416 | NP_056391.1 |
| NR_132749.1 | 19 | 5603 | ||
| XM_017006138.1 | 21 | 5682 | XP_016861627.1 | |
| XM_017006139.1 | 20 | 5632 | XP_016861628.1 | |
| XM_017006140.1 | 20 | 5610 | XP_016861629.1 | |
| XM_017006141.1 | 19 | 5622 | XP_016861630.1 | |
| XM_017006142.1 | 19 | 5601 | XP_016861631.1 | |
| XM_017006143.1 | 21 | 5676 | XP_016861632.1 | |
| XM_017006144.1 | 21 | 5705 | XP_016861633.1 | |
| XM_017006145.1 | 19 | 5574 | XP_016861634.1 | |
| XM_017006146.1 | 18 | 5586 | XP_016861635.1 | |
| XM_017006147.1 | 20 | 5693 | XP_016861636.1 | |
| XM_017006148.1 | 17 | 5574 | XP_016861637.1 | |
| XM_017006149.1 | 20 | 5726 | XP_016861638.1 | |
| XM_017006150.1 | 19 | 5480 | XP_016861639.1 | |
| XM_017006151.1 | 18 | 5565 | XP_016861640.1 | |
| XM_017006152.1 | 20 | 5640 | XP_016861641.1 | |
| XM_017006153.1 | 20 | 5669 | XP_016861642.1 | |
| XM_017006154.1 | 19 | 5453 | XP_016861643.1 | |
| XM_017006155.1 | 19 | 5737 | XP_016861644.1 | |
| XM_017006156.1 | 17 | 5553 | XP_016861645.1 | |
| XM_017006157.1 | 19 | 5628 | XP_016861646.1 | |
| XM_017006158.1 | 19 | 5660 | XP_016861647.1 | |
| XM_017006159.1 | 18 | 5687 | XP_016861648.1 | |
| AB002376.2 | 18 | 5416 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759150 | Chr.3:55456601 - 55631765 on Build GRCh38 | Loss |
|
| nsv517851 | Chr.3:54375499 - 55705332 on Build GRCh38 | Gain |
|
| nsv1013405 | Chr.3:55040452 - 56437132 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map