Assay Details
Target Gene Details
Entrez Gene ID: | 617 |
Gene Name: | BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone |
Gene Aliases: |
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1 |
Location: |
Chr.2:218659656-218663443 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCS1L | NM_001079866.1 | 8 | 1331 | NP_001073335.1 |
NM_001257342.1 | 9 | 1461 | NP_001244271.1 | |
NM_001257343.1 | 9 | 1513 | NP_001244272.1 | |
NM_001257344.1 | 8 | 1388 | NP_001244273.1 | |
NM_001318836.1 | 7 | 962 | NP_001305765.1 | |
NM_001320717.1 | 9 | 1503 | NP_001307646.1 | |
NM_004328.4 | 9 | 1540 | NP_004319.1 | |
XM_005246748.2 | 8 | 1815 | XP_005246805.1 | |
XM_005246749.3 | 6 | 814 | XP_005246806.1 | |
XM_006712678.1 | 8 | 1946 | XP_006712741.1 | |
XM_017004631.1 | 9 | 1702 | XP_016860120.1 | |
XM_017004632.1 | 9 | 1427 | XP_016860121.1 | |
XM_017004633.1 | 8 | 2205 | XP_016860122.1 | |
XM_017004634.1 | 7 | 1233 | XP_016860123.1 | |
AF026849.1 | 8 | 1307 | AAD08638.1 | |
AF038195.1 | 8 | 1269 | AAB97365.1 | |
AK096210.1 | 8 | 1894 | ||
AK223518.1 | 8 | 1417 | BAD97238.1 | |
AK289363.1 | 9 | 1403 | ||
BC000416.2 | 8 | 1461 | AAH00416.1 | |
BC007500.2 | 8 | 1461 | AAH07500.1 | |
BX571752.1 | 9 | 1460 | CAE11877.1 | |
KP992846.1 | 9 | 1460 | ||
KU177941.1 | 7 | 1194 | ||
KU177942.1 | 6 | 1049 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
