Assay Details
Target Gene Details
Entrez Gene ID: | 80204 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
FBX11, PRMT9, UBR6, UG063H01, VIT1 |
Location: |
Chr.2:47806920-47905793 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO11 | NM_001190274.1 | 23 | 3607 | NP_001177203.1 |
| NM_025133.4 | 23 | 3396 | NP_079409.3 | |
| XM_005264572.4 | 22 | 3682 | XP_005264629.1 | |
| XM_005264573.4 | 22 | 3635 | XP_005264630.1 | |
| XM_017005015.1 | 23 | 3554 | XP_016860504.1 | |
| XM_017005016.1 | 22 | 3501 | XP_016860505.1 | |
| XM_017005017.1 | 23 | 3420 | XP_016860506.1 | |
| AF264714.1 | 4 | 2623 | AAF76888.1 | |
| AI879484.1 | ||||
| AK025477.1 | 8 | 1599 | BAB15143.1 | |
| AL117620.1 | 6 | 1378 | CAB56019.1 | |
| BC043258.1 | 23 | 3498 | AAH43258.2 | |
| BC130445.1 | ||||
| BC136480.1 |
More Information
Set Membership: |
|

Genomic Map