Assay Details
Target Gene Details
Entrez Gene ID: | 79645 |
Gene Name: | EF-hand calcium binding domain 1 |
Gene Aliases: |
- |
Location: |
Chr.8:48714915-48735843 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB1 | NM_001142857.1 | NP_001136329.1 | ||
NM_024593.3 | 6 | 1750 | NP_078869.1 | |
NR_024605.1 | 6 | 1466 | ||
XM_005251303.1 | 7 | 1626 | XP_005251360.1 | |
XM_011517589.1 | XP_011515891.1 | |||
XM_011517590.2 | 7 | 1880 | XP_011515892.1 | |
XM_011517591.2 | 7 | 1853 | XP_011515893.1 | |
XM_017013826.1 | 6 | 1527 | XP_016869315.1 | |
XM_017013827.1 | 8 | 2003 | XP_016869316.1 | |
AK021829.1 | BAB13906.1 | |||
AK299659.1 | ||||
AK307101.1 | ||||
BC025676.1 | 6 | 1671 | AAH25676.1 | |
CB250588.1 | 1 | 386 | ||
DB536362.1 | 1 | 68 | ||
DB552638.1 | 1 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
