Assay Details
Target Gene Details
Entrez Gene ID: | 9318 |
Gene Name: | COP9 signalosome subunit 2 |
Gene Aliases: |
ALIEN, CSN2, SGN2, TRIP15 |
Location: |
Chr.15:49125274-49155657 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS2 | NM_001143887.1 | 13 | 1528 | NP_001137359.1 |
| NM_004236.3 | 13 | 1507 | NP_004227.1 | |
| AB209799.1 | 13 | 1454 | BAD93036.1 | |
| AF084260.1 | 13 | 1443 | AAC34122.1 | |
| AF100762.1 | 13 | 1433 | AAD43026.1 | |
| AF120268.1 | 13 | 1418 | AAD30269.1 | |
| AF212227.1 | 13 | 1454 | AAK26250.1 | |
| AK222590.1 | 13 | 1431 | BAD96310.1 | |
| AK295598.1 | 11 | 1262 | ||
| BC012629.1 | 13 | 1444 | AAH12629.1 |
More Information
Set Membership: |
|

Genomic Map