Assay Details
Target Gene Details
Entrez Gene ID: | 51654 |
Gene Name: | CDK5 regulatory subunit associated protein 1 |
Gene Aliases: |
C20orf34, C42, CGI-05, HSPC167 |
Location: |
Chr.20:33358839-33402768 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map