Assay Details
Target Gene Details
Entrez Gene ID: | 55904 |
Gene Name: | lysine methyltransferase 2E |
Gene Aliases: |
HDCMC04P, MLL5, NKp44L |
Location: |
Chr.7:105014190-105114085 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT2E | NM_018682.3 | NP_061152.3 | ||
| NM_182931.2 | NP_891847.1 | |||
| XM_005250493.1 | XP_005250550.1 | |||
| XM_011516400.1 | XP_011514702.1 | |||
| XM_017012435.1 | XP_016867924.1 | |||
| AF067804.1 | AAF75564.1 | |||
| AF519459.1 | AAM74947.1 | |||
| AK000940.1 | ||||
| AY147037.1 | AAN17675.1 | |||
| AY157990.1 | AAN76325.1 | |||
| AY211525.1 | AAO86416.1 | |||
| BC001296.2 | AAH01296.1 | |||
| BC062583.1 | AAH62583.1 | |||
| BC142987.1 | ||||
| JQ809698.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608068 | Chr.7:104981955 - 105117036 on Build GRCh38 | Loss |
|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map