Assay Details
Target Gene Details
Entrez Gene ID: | 55167 |
Gene Name: | male-specific lethal 2 homolog (Drosophila) |
Gene Aliases: |
MSL-2, MSL2L1, RNF184 |
Location: |
Chr.3:136148918-136197300 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSL2 | NM_001145417.1 | 2 | 437 | NP_001138889.1 |
| NM_018133.3 | 2 | 1238 | NP_060603.2 | |
| XM_005247571.3 | 2 | 972 | XP_005247628.1 | |
| XM_006713684.3 | 2 | 455 | XP_006713747.1 | |
| XM_011512949.2 | 2 | 542 | XP_011511251.1 | |
| AB046805.1 | 2 | 586 | ||
| AK091956.1 | 2 | 1238 | ||
| AK292557.1 | 2 | 733 | ||
| AK302491.1 | 2 | 404 | ||
| AL834289.1 | 2 | 556 | CAD38963.2 | |
| BC032719.1 | ||||
| BC093764.1 | 2 | 538 | AAH93764.1 | |
| BC093790.1 | 2 | 537 | AAH93790.1 | |
| DC409643.1 | 2 | 523 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4024 | Chr.3:136146065 - 136158844 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map