Assay Details
Target Gene Details
Entrez Gene ID: | 102723833 |
Gene Name: | uncharacterized LOC102723833 |
Gene Aliases: |
- |
Location: |
Chr.1:216072465-216086917 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723833 | NR_125992.1 | |||
| NR_125993.1 | ||||
| AI201399.1 | ||||
| AW119097.1 | ||||
| BX340080.2 |
Target Gene Details
Entrez Gene ID: | 7399 |
Gene Name: | usherin |
Gene Aliases: |
RP39, US2, USH2, dJ1111A8.1 |
Location: |
Chr.1:215622894-216423396 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USH2A | NM_206933.2 | NP_996816.2 | ||
| AY481573.1 | AAS47698.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74766738] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map