Assay Details
Target Gene Details
Entrez Gene ID: | 25949 |
Gene Name: | SYF2 pre-mRNA splicing factor |
Gene Aliases: |
CBPIN, NTC31, P29, fSAP29 |
Location: |
Chr.1:25222276-25232522 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYF2 | NM_015484.4 | 7 | 1293 | NP_056299.1 |
| NM_207170.3 | 6 | 1167 | NP_997053.1 | |
| AF273089.1 | 7 | 1253 | AAG42073.1 | |
| AL080166.1 | 7 | 1254 | CAB45754.1 | |
| BC010862.1 | 7 | 1264 | AAH10862.1 | |
| BF057608.1 | 1 | 423 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv132n100 | Chr.1:24981181 - 25393108 on Build GRCh38 | Gain |
|
| esv3576582 | Chr.1:24981195 - 25361084 on Build GRCh38 | Gain |
|
| nsv7187 | Chr.1:25197127 - 25229506 on Build GRCh38 | Insertion |
|
| nsv1160388 | Chr.1:25177805 - 25266637 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map