Assay Details
Target Gene Details
Entrez Gene ID: | 6432 |
Gene Name: | serine and arginine rich splicing factor 7 |
Gene Aliases: |
9G8, AAG3, SFRS7 |
Location: |
Chr.2:38743599-38751494 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF7 | NM_001031684.2 | 3 | 570 | NP_001026854.1 |
| NM_001195446.1 | 3 | 570 | NP_001182375.1 | |
| XM_005264484.2 | 3 | 437 | XP_005264541.1 | |
| XM_005264485.2 | 3 | 437 | XP_005264542.1 | |
| AB445102.1 | 3 | 332 | ||
| AK091425.1 | 3 | 385 | BAC03661.1 | |
| AK225141.1 | 3 | 437 | ||
| AK293667.1 | 3 | 437 | ||
| AK297161.1 | 3 | 437 | ||
| AY166860.1 | 3 | 458 | AAN87842.1 | |
| AY513287.1 | 3 | 413 | AAT08040.1 | |
| BC000997.2 | 3 | 417 | AAH00997.1 | |
| BC017369.2 | 3 | 428 | AAH17369.1 | |
| BC017908.1 | 3 | 435 | AAH17908.1 | |
| BC022328.1 | 3 | 435 | AAH22328.1 | |
| BT006745.1 | 3 | 332 | AAP35391.1 | |
| DC358626.1 | ||||
| L22253.1 | 3 | 437 | AAA35495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763640 | Chr.2:38727277 - 38750229 on Build GRCh38 | Gain |
|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map