Assay Details
Target Gene Details
Entrez Gene ID: | 23522 |
Gene Name: | lysine acetyltransferase 6B |
Gene Aliases: |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
Location: |
Chr.10:74824927-75032623 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551535 | Chr.10:74745813 - 74842047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map