Assay Details
Target Gene Details
Entrez Gene ID: | 5264 |
Gene Name: | phytanoyl-CoA 2-hydroxylase |
Gene Aliases: |
LN1, LNAP1, PAHX, PHYH1, RD |
Location: |
Chr.10:13277796-13300130 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYH | NM_001037537.1 | NP_001032626.1 | ||
| NM_001323080.1 | NP_001310009.1 | |||
| NM_001323082.1 | NP_001310011.1 | |||
| NM_001323083.1 | NP_001310012.1 | |||
| NM_001323084.1 | NP_001310013.1 | |||
| NM_006214.3 | NP_006205.1 | |||
| AF023462.1 | AAB81834.1 | |||
| AF112977.1 | AAD20602.1 | |||
| BC029512.1 | AAH29512.1 | |||
| BF509272.1 | ||||
| CR542055.1 | CAG46852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831788 | Chr.10:13291241 - 13469810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map