Assay Details
Target Gene Details
Entrez Gene ID: | 9612 |
Gene Name: | nuclear receptor corepressor 2 |
Gene Aliases: |
CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, TNRC14, TRAC, TRAC-1, TRAC1 |
Location: |
Chr.12:124324411-124567464 on Build GRCh38 |
Assay Gene Location: | Within Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOR2 | NM_001077261.3 | 48 | 8125 | NP_001070729.2 |
| NM_001206654.1 | 48 | 8263 | NP_001193583.1 | |
| NM_006312.5 | 49 | 8293 | NP_006303.4 | |
| AF113003.1 | 48 | 7982 | AAD20946.1 | |
| AF125672.1 | 47 | 8107 | AAD22973.1 | |
| AY965853.1 | 47 | 7969 | AAX77219.1 | |
| BC004326.1 | 10 | 2248 | AAH04326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818930 | Chr.12:124256444 - 124391587 on Build GRCh38 | Gain |
|
| esv3631012 | Chr.12:124290695 - 124349764 on Build GRCh38 | Gain |
|
| esv3631010 | Chr.12:124288701 - 124749461 on Build GRCh38 | Gain |
|
| nsv952500 | Chr.12:124296455 - 124567454 on Build GRCh38 | Deletion |
|
| esv3631011 | Chr.12:124289768 - 124548418 on Build GRCh38 | Gain |
|
| nsv470322 | Chr.12:124313867 - 124452057 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map