Assay Details
Target Gene Details
Entrez Gene ID: | 27349 |
Gene Name: | malonyl-CoA-acyl carrier protein transacylase |
Gene Aliases: |
FASN2C, MCT, MT, NET62, fabD |
Location: |
Chr.22:43132206-43143397 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCAT | NM_014507.3 | 1 | 349 | NP_055322.1 |
| NM_173467.4 | 1 | 349 | NP_775738.3 | |
| NR_046423.1 | 1 | 349 | ||
| AK314059.1 | 1 | 331 | ||
| AL359401.1 | 1 | 316 | CAB94789.1 | |
| AL359403.1 | 1 | 316 | CAB94790.1 | |
| BC030985.1 | 1 | 301 | AAH30985.1 | |
| BC042195.1 | 1 | 342 | AAH42195.2 | |
| BI464466.1 | 1 | 355 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762124 | Chr.22:43031130 - 43188670 on Build GRCh38 | Gain |
|
| esv33816 | Chr.22:43113758 - 43326122 on Build GRCh38 | Gain |
|
| nsv834214 | Chr.22:43037931 - 43217928 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map