Assay Details
Target Gene Details
Entrez Gene ID: | 254783 |
Gene Name: | olfactory receptor family 6 subfamily C member 74 |
Gene Aliases: |
- |
Location: |
Chr.12:55244114-55248226 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6C74 | NM_001005490.1 | 1 | 567 | NP_001005490.1 |
| XM_017019126.1 | 2 | 648 | XP_016874615.1 | |
| BC140754.1 | 1 | 657 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map