Assay Details
Target Gene Details
Entrez Gene ID: | 6699 |
Gene Name: | small proline rich protein 1B |
Gene Aliases: |
CORNIFIN, GADD33, SPR-IB, SPRR1 |
Location: |
Chr.1:153031203-153032900 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRR1B | NM_003125.2 | 2 | 546 | NP_003116.2 |
| BC056240.1 | 2 | 550 | AAH56240.1 | |
| CK300778.1 | 1 | 91 | ||
| M19888.1 | 2 | 550 | AAA36636.1 | |
| M21300.1 | 2 | 546 | AAA36637.1 | |
| M21301.1 | 2 | 546 | AAA36638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map