Assay Details
Target Gene Details
Entrez Gene ID: | 339479 |
Gene Name: | BMP/retinoic acid inducible neural specific 3 |
Gene Aliases: |
DBCCR1L, DBCCR1L1, FAM5C |
Location: |
Chr.1:190097662-190477997 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRINP3 | NM_001317188.1 | 7 | 1383 | NP_001304117.1 |
| NM_199051.2 | 8 | 1778 | NP_950252.1 | |
| XM_011509472.2 | 9 | 1780 | XP_011507774.1 | |
| XM_011509475.2 | 8 | 1518 | XP_011507777.1 | |
| XM_011509476.2 | 7 | 1596 | XP_011507778.1 | |
| XM_017001125.1 | 10 | 1889 | XP_016856614.1 | |
| XM_017001126.1 | 9 | 1754 | XP_016856615.1 | |
| XM_017001127.1 | 9 | 1918 | XP_016856616.1 | |
| XM_017001128.1 | 9 | 3006 | XP_016856617.1 | |
| XM_017001129.1 | 7 | 1587 | XP_016856618.1 | |
| XM_017001130.1 | 8 | 1563 | XP_016856619.1 | |
| XM_017001131.1 | 9 | 1868 | XP_016856620.1 | |
| AB111893.1 | 8 | 1525 | BAD04066.1 | |
| AK123024.1 | 8 | 1468 | ||
| AK123637.1 | 8 | 1523 | ||
| AK294365.1 | 7 | 1185 | ||
| BC105052.1 | 7 | 1349 | AAI05053.1 | |
| BC105054.1 | 7 | 1349 | AAI05055.1 | |
| U67037.1 | 2 | 324 | AAD09521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762036 | Chr.1:190073576 - 190122891 on Build GRCh38 | Gain |
|
| dgv693n54 | Chr.1:190067010 - 190110606 on Build GRCh38 | Loss |
|
| nsv832104 | Chr.1:190021018 - 190217578 on Build GRCh38 | Gain |
|
| dgv694n54 | Chr.1:190094081 - 190147232 on Build GRCh38 | Loss |
|
| nsv548532 | Chr.1:189789642 - 190402541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map