Assay Details
Target Gene Details
Entrez Gene ID: | 27306 |
Gene Name: | hematopoietic prostaglandin D synthase |
Gene Aliases: |
GSTS, GSTS1, GSTS1-1, PGD2, PGDS |
Location: |
Chr.4:94298556-94342876 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPGDS | NM_014485.2 | 2 | 126 | NP_055300.1 |
| XM_005262932.2 | 2 | 126 | XP_005262989.1 | |
| AK290075.1 | 2 | 126 | ||
| BC020734.1 | 2 | 103 | AAH20734.1 | |
| CR541662.1 | CAG46463.1 | |||
| CR541679.1 | CAG46480.1 | |||
| D82073.1 | 1 | 45 | BAA25545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594901 | Chr.4:94333579 - 94572095 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148378201] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map