Assay Details
Target Gene Details
Entrez Gene ID: | 28962 |
Gene Name: | osteopetrosis associated transmembrane protein 1 |
Gene Aliases: |
GIPN, GL, HSPC019, OPTB5 |
Location: |
Chr.6:108041409-108074737 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSTM1 | NM_014028.3 | 6 | 3835 | NP_054747.2 |
| AJ420489.1 | 2 | 1903 | ||
| BE218267.1 | 1 | 227 | ||
| BQ716123.1 | 1 | 502 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1031826 | Chr.6:107969391 - 108109876 on Build GRCh38 | Loss |
|
| nsv604456 | Chr.6:107999992 - 108043747 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map