Assay Details
Target Gene Details
Entrez Gene ID: | 8200 |
Gene Name: | growth differentiation factor 5 |
Gene Aliases: |
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2 |
Location: |
Chr.20:35433347-35454746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF5 | NM_000557.4 | NP_000548.2 | ||
| NM_001319138.1 | NP_001306067.1 | |||
| XM_011529075.2 | XP_011527377.1 | |||
| AB593125.1 | ||||
| BC032495.1 | AAH32495.1 | |||
| U13660.1 | AAA57007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833963 | Chr.20:35330383 - 35498678 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map