Assay Details
Target Gene Details
Entrez Gene ID: | 29881 |
Gene Name: | NPC1 like intracellular cholesterol transporter 1 |
Gene Aliases: |
NPC11L1 |
Location: |
Chr.7:44512440-44541650 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPC1L1 | NM_001101648.1 | NP_001095118.1 | ||
| NM_013389.2 | NP_037521.2 | |||
| XM_011515326.2 | XP_011513628.1 | |||
| XM_011515328.2 | XP_011513630.1 | |||
| AF192522.1 | AAF20396.1 | |||
| AY437865.1 | AAR97886.1 | |||
| AY515256.1 | AAS56939.1 | |||
| AY927603.1 | ||||
| BC117178.1 | ||||
| BC143756.1 | ||||
| FJ481111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523191 | Chr.7:44513639 - 44541392 on Build GRCh38 | Loss |
|
| nsv606806 | Chr.7:44427130 - 44541392 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map