Assay Details
Target Gene Details
Entrez Gene ID: | 5784 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 14 |
Gene Aliases: |
PEZ, PTP36 |
Location: |
Chr.1:214348696-214551681 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN14 | NM_005401.4 | 19 | 4524 | NP_005392.2 |
| XM_017001941.1 | 20 | 4611 | XP_016857430.1 | |
| AW192897.1 |
More Information
Set Membership: |
|

Genomic Map