Assay Details
Target Gene Details
Entrez Gene ID: | 57129 |
Gene Name: | mitochondrial ribosomal protein L47 |
Gene Aliases: |
CGI-204, L47mt, MRP-L47, NCM1 |
Location: |
Chr.3:179588465-179604646 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL47 | NM_020409.2 | 7 | 1075 | NP_065142.2 |
| NM_177988.1 | 6 | 929 | NP_817125.1 | |
| BC032522.1 | 7 | 1008 | AAH32522.1 |
More Information
Set Membership: |
|

Genomic Map