Assay Details
Target Gene Details
Entrez Gene ID: | 9443 |
Gene Name: | mediator complex subunit 7 |
Gene Aliases: |
ARC34, CRSP33, CRSP9 |
Location: |
Chr.5:157138440-157142910 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED7 | NM_001100816.1 | 2 | 562 | NP_001094286.1 |
| NM_004270.4 | 2 | 278 | NP_004261.1 | |
| AF031383.1 | 2 | 236 | AAC52115.1 | |
| AF104251.1 | 1 | 170 | AAD12720.1 | |
| AK308393.1 | 2 | 243 | ||
| AV711090.1 | 2 | 290 | ||
| BT007368.1 | 1 | 170 | AAP36032.1 | |
| CR457009.1 | 1 | 170 | CAG33290.1 | |
| CR541696.1 | 1 | 170 | CAG46497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map