Assay Details
Target Gene Details
Entrez Gene ID: | 5905 |
Gene Name: | Ran GTPase activating protein 1 |
Gene Aliases: |
Fug1, RANGAP, SD |
Location: |
Chr.22:41244777-41302213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RANGAP1 | NM_001317930.1 | NP_001304859.1 | ||
BC041396.2 | AAH41396.1 |
Target Gene Details
Entrez Gene ID: | 23264 |
Gene Name: | zinc finger CCCH-type containing 7B |
Gene Aliases: |
RoXaN |
Location: |
Chr.22:41301503-41360147 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZC3H7B | NM_017590.5 | 1 | 153 | NP_060060.3 |
XM_011530016.2 | 1 | 134 | XP_011528318.1 | |
AF188530.1 | AAF05541.1 | |||
AK023849.1 | ||||
AK225891.1 | 1 | 126 | ||
AK315009.1 | 1 | 94 | ||
BC152558.1 | 1 | 131 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv3638 | Chr.22:41285245 - 41309688 on Build GRCh38 | Insertion |
![]() ![]() |
dgv772n67 | Chr.22:41290675 - 41308013 on Build GRCh38 | Gain |
![]() ![]() |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829242 | Chr.22:41301601 - 41303153 on Build GRCh38 | Gain |
![]() ![]() |
nsv817997 | Chr.22:41244293 - 41366866 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
