Assay Details
Target Gene Details
Entrez Gene ID: | 27231 |
Gene Name: | nicotinamide riboside kinase 2 |
Gene Aliases: |
ITGB1BP3, MIBP, NRK2 |
Location: |
Chr.19:3933103-3942416 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMRK2 | NM_001289117.1 | 2 | 268 | NP_001276046.1 |
| NM_170678.2 | 2 | 268 | NP_733778.1 | |
| NR_110316.1 | ||||
| XM_006722725.3 | 1 | 179 | XP_006722788.1 | |
| XM_006722726.3 | 1 | 176 | XP_006722789.1 | |
| XM_017026597.1 | 1 | 171 | XP_016882086.1 | |
| AF190819.1 | 1 | 51 | AAF26711.1 | |
| AK001663.1 | 2 | 278 | BAA91820.1 | |
| AK022514.1 | 1 | 110 | BAB14071.1 | |
| AL365377.1 | CAB96949.1 | |||
| AY611481.1 | AAT11929.1 | |||
| BC093637.1 | AAH93637.1 | |||
| BC101575.1 | AAI01576.1 | |||
| BC143329.1 | ||||
| BM717225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833715 | Chr.19:3786758 - 3977144 on Build GRCh38 | Loss |
|
| esv3643483 | Chr.19:3932245 - 3944820 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map